Anti-DES Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3221-02AEA 4050 DKK
USBID3221-02A
Anti-DES Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:50-1:100

Type: Primary
Antigen: DES (desmin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mammalian
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR