Anti-DES Mouse monoclonal antibody [clone: 6A64]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3221-01AEA 3140 DKK
USBID3221-01A
Anti-DES Mouse monoclonal antibody [clone: 6A64]
Antistoffer
Type: Primary
Antigen: DES (desmin)
Clonality: monoclonal
Clone: 6A64
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR