Anti-DCD Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3213-01CEA 3700 DKK
USBID3213-01C
Anti-DCD Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:50-1:100; Western Blot: 1:1000; Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: DCD (dermcidin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR