Anti-DERA Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3206-99-APEA 6360 DKK
USBID3206-99-AP
Anti-DERA Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: DERA (deoxyribose-phosphate aldolase (putative))
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR