Anti-DEPDC4 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3206-92EA 3140 DKK
USBID3206-92
Anti-DEPDC4 Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1: 8000

Type: Primary
Antigen: DEPDC4 (DEP domain containing 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR