Anti-DUT Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10C14]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3206-04-PEEA 0 DKK
USBID3206-04-PE
Anti-DUT Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10C14]
Type: Primary
Antigen: DUT (deoxyuridine triphosphatase)
Clonality: monoclonal
Clone: 10C14
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR