Anti-DUT Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3206-04AEA 3290 DKK
USBID3206-04A
Anti-DUT Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry:1-2?g/ml

Type: Primary
Antigen: DUT (deoxyuridine triphosphatase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR