Anti-Deoxyribonuclease II Rabbit polyclonal antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID3201-22BEA 5630 DKK
USBID3201-22B
Anti-Deoxyribonuclease II Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: Deoxyribonuclease II
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human