Anti-DNASE1 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3200-05EEA 3940 DKK
USBID3200-05E
Anti-DNASE1 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:500-1:1500

Type: Primary
Antigen: DNASE1 (deoxyribonuclease I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR