Anti-DGUOK Rabbit Polyclonal Antibody (Biotin)
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID3188-10A-BIOTEA 6360 DKK
USBID3188-10A-BIOT
Anti-DGUOK Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: DGUOK (deoxyguanosine kinase)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse