Anti-DCK Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3185-25BEA 4320 DKK
USBID3185-25B
Anti-DCK Rabbit polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1 ?g/ml; Western Blot: 1 ?g/ml; Immunochemistry: 20 ?g/ml

Type: Primary
Antigen: DCK (deoxycytidine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR