Anti-TSPYL2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3155-01EA 3180 DKK
USBID3155-01
Anti-TSPYL2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TSPYL2 (TSPY-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR