Anti-Density Regulated Protein Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2711]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3154-01-FITCEA 0 DKK
USBID3154-01-FITC
Anti-Density Regulated Protein Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2711]
Type: Primary
Antigen: Density Regulated Protein
Clonality: monoclonal
Clone: 10B2711
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR