Anti-DENND4C Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3150-40EA 3460 DKK
USBID3150-40
Anti-DENND4C Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:8000; Western Blot: 0.5-2?g/ml

Type: Primary
Antigen: DENND4C (DENN/MADD domain containing 4C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR