Anti-Dengue Types 1,2,3,4 Mouse monoclonal antibody [clone: 9L151]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID2800-25AEA 8290 DKK
USBID2800-25A
Anti-Dengue Types 1,2,3,4 Mouse monoclonal antibody [clone: 9L151]
Antistoffer
Type: Primary
Antigen: Dengue Types 1,2,3,4
Clonality: monoclonal
Clone: 9L151
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR