Anti-Dengue Type 2 Mouse monoclonal antibody [clone: 5C36]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID2800-14EA 3230 DKK
USBID2800-14
Anti-Dengue Type 2 Mouse monoclonal antibody [clone: 5C36]
Antistoffer
Recommended dilution: Immunochemistry: 1:1000

Type: Primary
Antigen: Dengue Type 2
Clonality: monoclonal
Clone: 5C36
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR