Anti-ECI1 Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID2744-90-BIOTEA 6360 DKK
USBID2744-90-BIOT
Anti-ECI1 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: ECI1 (enoyl-CoA delta isomerase 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR