Anti-Dendritic Cell Immunoactivating Receptor Rat monoclonal antibody [clone: 7H115]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID2744-55EA 4320 DKK
USBID2744-55
Anti-Dendritic Cell Immunoactivating Receptor Rat monoclonal antibody [clone: 7H115]
Antistoffer
Type: Primary
Antigen: Dendritic Cell Immunoactivating Receptor
Clonality: monoclonal
Clone: 7H115
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR