Anti-Dll-4 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID2731-08EA 2900 DKK
USBID2731-08
Anti-Dll-4 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:5000-1:20000

Type: Primary
Antigen: Dll-4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR