Anti-Delta 4 Rat monoclonal antibody [clone: 9H111]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID2728-01CEA 7310 DKK
USBID2728-01C
Anti-Delta 4 Rat monoclonal antibody [clone: 9H111]
Antistoffer
Recommended dilution: ELISA: 2-8?g/ml
Type: Primary
Antigen: Delta 4
Clonality: monoclonal
Clone: 9H111
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse