Anti-Delta 4 Rat Monoclonal Antibody (Biotin) [clone: 9H110]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID2728-01BEA 6790 DKK
USBID2728-01B
Anti-Delta 4 Rat Monoclonal Antibody (Biotin) [clone: 9H110]
Antistoffer
Recommended dilution: ELISA: 0.5-2?g/ml

Type: Primary
Antigen: Delta 4
Clonality: monoclonal
Clone: 9H110
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR