Anti-Delta 4 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID2728EA 5010 DKK
USBID2728
Anti-Delta 4 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:5000-1:25000; Immunochemistry: 10-20 ?g/ml; Western Blot: 1:500-1:2500

Type: Primary
Antigen: Delta 4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR