Anti-DLL3 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID2727-85AEA 4030 DKK
USBID2727-85A
Anti-DLL3 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 3-5 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: DLL3 (delta-like 3 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR