Anti-DLL3 Mouse monoclonal antibody [clone: 7H171]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID2727-85EA 4590 DKK
USBID2727-85
Anti-DLL3 Mouse monoclonal antibody [clone: 7H171]
Antistoffer
Recommended dilution: Western Blot: 1-2?g/ml

Type: Primary
Antigen: DLL3 (delta-like 3 (Drosophila))
Clonality: monoclonal
Clone: 7H171
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR