Anti-DLL1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 12K77]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID2727-75EEA 4140 DKK
USBID2727-75E
Anti-DLL1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 12K77]
Antistoffer
Do not freeze!

Type: Primary
Antigen: DLL1 (delta-like 1 (Drosophila))
Clonality: monoclonal
Clone: 12K77
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR