Anti-DLL1 Hamster monoclonal antibody [clone: 12G25]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID2727-75CEA 4440 DKK
USBID2727-75C
Anti-DLL1 Hamster monoclonal antibody [clone: 12G25]
Antistoffer
Type: Primary
Antigen: DLL1 (delta-like 1 (Drosophila))
Clonality: monoclonal
Clone: 12G25
Conjugation: unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR