Anti-Delta-like 1 Mouse monoclonal antibody [clone: 7H56]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID2727-75EA 4320 DKK
USBID2727-75
Anti-Delta-like 1 Mouse monoclonal antibody [clone: 7H56]
Antistoffer
Recommended dilution: Immunochemistry: 8-25 ?g/ml

Type: Primary
Antigen: Delta-like 1
Clonality: monoclonal
Clone: 7H56
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR