Anti-Delta-like 1 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID2727-70DEA 3460 DKK
USBID2727-70D
Anti-Delta-like 1 Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:16000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: Delta-like 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR