Anti-Delta-like 1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9H109]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID2727-70CEA 5560 DKK
USBID2727-70C
Anti-Delta-like 1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9H109]
Antistoffer
Do not freeze!

Type: Primary
Antigen: Delta-like 1
Clonality: monoclonal
Clone: 9H109
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR