Anti-Delta-like 1 Sheep polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID2727-70BEA 5920 DKK
USBID2727-70B
Anti-Delta-like 1 Sheep polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 0.5-1.0 ?g/ml; Western Blot: 0.1-0.2?g/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: Delta-like 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR