Anti-DAZL Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID2675-02BEA 3180 DKK
USBID2675-02B
Anti-DAZL Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:5000

Type: Primary
Antigen: DAZL (deleted in azoospermia-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Human, Monkey, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR