Anti-DAZ1 Mouse monoclonal antibody [clone: 3G10]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID2674-65CEA 4790 DKK
USBID2674-65C
Anti-DAZ1 Mouse monoclonal antibody [clone: 3G10]
Antistoffer
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: DAZ1 (deleted in azoospermia 1)
Clonality: monoclonal
Clone: 3G10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR