Anti-DEAF1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1957-90AEA 3700 DKK
USBID1957-90A
Anti-DEAF1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: DEAF1 (DEAF1 transcription factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR