Anti-DEDD2 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1890-02CEA 3270 DKK
USBID1890-02C
Anti-DEDD2 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000-1:2000; Immunochemistry: 1:1000-1:5000; Immunoprecipitation: 1:50-1:200

Type: Primary
Antigen: DEDD2 (death effector domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR