Anti-DEDD2 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1890-02EA 3340 DKK
USBID1890-02
Anti-DEDD2 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 05-1 ?g/ml

Type: Primary
Antigen: DEDD2 (death effector domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR