Anti-RYBP Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1882-01CEA 4040 DKK
USBID1882-01C
Anti-RYBP Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 0.5-1.0 ?g/ml

Type: Primary
Antigen: RYBP (RING1 and YY1 binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR