Anti-Defender of Apoptotic Cell Death 1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1877-15BEA 4940 DKK
USBID1877-15B
Anti-Defender of Apoptotic Cell Death 1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 0.5-2.0 ?g/ml

Type: Primary
Antigen: Defender of Apoptotic Cell Death 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR