Anti-DECR1 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2703]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1876-95-APEA 0 DKK
USBID1876-95-AP
Anti-DECR1 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2703]
Type: Primary
Antigen: DECR1 (2,4-dienoyl CoA reductase 1, mitochondrial)
Clonality: monoclonal
Clone: 10B2703
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR