Anti-Dectin 2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10B2695]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1876-49D7EA 3230 DKK
USBID1876-49D7
Anti-Dectin 2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10B2695]
Antistoffer
Recommended dilution: Flow Cytometry:1:5;
Do not freeze!

Type: Primary
Antigen: Dectin 2 (dectin 2)
Clonality: monoclonal
Clone: 10B2695
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR