Anti-Dectin 2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 9L823]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1876-49CEA 4320 DKK
USBID1876-49C
Anti-Dectin 2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 9L823]
Antistoffer
Do not freeze!

Type: Primary
Antigen: Dectin 2 (dectin 2)
Clonality: monoclonal
Clone: 9L823
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR