Anti-Dectin 2 Mouse monoclonal antibody Carboxyfluorescein [clone: 9L822]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1876-49BEA 3680 DKK
USBID1876-49B
Anti-Dectin 2 Mouse monoclonal antibody Carboxyfluorescein [clone: 9L822]
Antistoffer
Type: Primary
Antigen: Dectin 2 (dectin 2)
Clonality: monoclonal
Clone: 9L822
Conjugation: Carboxyfluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR