Anti-Dectin-1 Rat monoclonal antibody [clone: 10B2689]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1876-48P2EA 4190 DKK
USBID1876-48P2
Anti-Dectin-1 Rat monoclonal antibody [clone: 10B2689]
Antistoffer
Recommended dilution: Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: Dectin-1
Clonality: monoclonal
Clone: 10B2689
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR