Anti-Dectin-1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 8B13]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID1876-48EEA 5440 DKK
USBID1876-48E
Anti-Dectin-1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 8B13]
Antistoffer
Do not freeze!
Type: Primary
Antigen: Dectin-1
Clonality: monoclonal
Clone: 8B13
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse