Anti-Dectin-1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 8B11]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1876-48CEA 4500 DKK
USBID1876-48C
Anti-Dectin-1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 8B11]
Antistoffer
Do not freeze!

Type: Primary
Antigen: Dectin-1
Clonality: monoclonal
Clone: 8B11
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR