Anti-Dectin-1, Low Endotoxin Mouse monoclonal antibody [clone: 9i100]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1876-48AEA 6500 DKK
USBID1876-48A
Anti-Dectin-1, Low Endotoxin Mouse monoclonal antibody [clone: 9i100]
Antistoffer
Recommended dilution: Flow Cytometry: 1:25

Type: Primary
Antigen: Dectin-1, Low Endotoxin
Clonality: monoclonal
Clone: 9i100
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR