Anti-DCN Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2685]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1875-07-APEA 0 DKK
USBID1875-07-AP
Anti-DCN Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2685]
Type: Primary
Antigen: DCN (decorin)
Clonality: monoclonal
Clone: 10B2685
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR