Anti-DCN Mouse monoclonal antibody [clone: 3H4-1F4]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1875-06DEA 4730 DKK
USBID1875-06D
Anti-DCN Mouse monoclonal antibody [clone: 3H4-1F4]
Antistoffer
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: DCN (decorin)
Clonality: monoclonal
Clone: 3H4-1F4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR