Anti-DCN Sheep polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1875-05CEA 4320 DKK
USBID1875-05C
Anti-DCN Sheep polyclonal antibody
Antistoffer
Recommended dilution: Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: DCN (decorin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR