Anti-DDX5 Mouse monoclonal antibody [clone: 8C102]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1660-40EA 2530 DKK
USBID1660-40
Anti-DDX5 Mouse monoclonal antibody [clone: 8C102]
Antistoffer
Recommended dilution: Western Blot: 0.2-2?g/ml; Immunoprecipitation: 100-500 ?g/sample; Immunochemistry: 2-100 ?g/ml

Type: Primary
Antigen: DDX5 (DEAD (Asp-Glu-Ala-Asp) box helicase 5)
Clonality: monoclonal
Clone: 8C102
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR