Anti-DDX4 Mouse monoclonal antibody [clone: 11C1025]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID1660-27EA 3350 DKK
USBID1660-27
Anti-DDX4 Mouse monoclonal antibody [clone: 11C1025]
Antistoffer
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50

Type: Primary
Antigen: DDX4 (DEAD (Asp-Glu-Ala-Asp) box polypeptide 4)
Clonality: monoclonal
Clone: 11C1025
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR